missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Kallikrein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534876
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: mouse kidney. Predicted reactivity: Dog (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plasma Kallikrein Polyclonal antibody specifically detects Plasma Kallikrein in Human, Mouse samples. It is validated for Western Blot.Spezifikation
Plasma Kallikrein | |
Polyclonal | |
Unconjugated | |
Klkb1 | |
antigen, prostate specific; APS; EC 3.4.21.34; Fletcher factor; Kal3; Kal-3; kallikrein; kallikrein 3, plasma; kallikrein B plasma; kallikrein B, plasma (Fletcher factor) 1; kallikrein B, plasma 1; kallikrein B1; Kalp1; KALP15; kininogenin; Klk3; KLKB1; Pk; PKK; PKKD; plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; PPK; prekallikrein; PSA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16621, 3818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P03952, P26262 | |
Klkb1 | |
Recombinant fragment corresponding to a region within amino acids 316 and 597 of Plasma Kallikrein 1B (Uniprot ID#P03952) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |