Learn More
PAI1 Goat anti-Human, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA186706
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with chimpanzee and dog based on sequence homology. Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is human placenta. In Western blot, this antibody detects a band at ∽45kDa in human duodenum, liver and lung lysates. A minimum incubation time of 1 hour is recommended with this antibody. Goat anti human PAI-1 antibody recognizes human Plasminogen activator inhibitor 1 (PAI-1), also known as Serpin E1.
PAI1 (plasminogen activator inhibitor 1) belongs to serine protease inhibitor superfamily, and is the principal inhibitor of tissue-type and urokinase-type plasminogen activators (tPA and uPA). Platelets are the main source of the circulating PAI11, but it is synthesized and secreted by many tissue and cell types, including fibroblasts, smooth muscle cells, endothelial cells, hepatocytes, and inflammatory cells. Expression of PAI11 can be regulated at the transcriptional level by many factors including growth factors, cytokines, hormones, inflammatory factors, glucose or lipid metabolites, vascular tone regulating factors, chemicals, and other environmental or physical factors. PAI1 is present at increased levels in various disease states, and has been linked to an increased occurrence of thrombosis in obesity, thrombophilia and the metabolic syndrome. Defects in PAI-1 are characterized by abnormal bleeding. PAI1 mediates inhibition of fibrinolysis by inhibiting the activity of plasminogen activator, and may promote neuronal survival. Other defects in PAI1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency). Alternatively spliced transcript variants encoding different isoforms of PAI1 have been found.Spezifikation
PAI1 | |
Polyclonal | |
Unconjugated | |
P05121 | |
SERPINE1 | |
Peptide with sequence C-GFKIDDKGMAPALRH, corresponding to the internal region of PAI-1 (NP_000593.1). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
SERPINE1 | |
PAI-1 | |
Goat | |
Affinity chromatography | |
RUO | |
5054 | |
4°C or -20°C if preferred | |
Liquid |