missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasminogen Mouse anti-Human, Clone: 10A1, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen LFMA0170
Weitere Details : Netto-Gewicht : 0.99000kg
Beschreibung
A suggested positive control for this product is human plasma.
The protein encoded by this gene is a secreted blood zymogen that is activated by proteolysis and converted to plasmin and angiostatin. Plasmin dissolves fibrin in blood clots and is an important protease in many other cellular processes while angiostatin inhibits angiogenesis. Defects in this gene are likely a cause of thrombophilia and ligneous conjunctivitis. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
Plasminogen | |
Monoclonal | |
Unconjugated | |
P00747 | |
PLG | |
Protein purified from Human plasma | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
10A1 | |
PLG | |
RP1-81D8.1, DKFZp779M0222 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
5340 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |