Learn More
Plasminogen Kringle 5 Rat anti-Mouse, Clone: 120304, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB742
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411766
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Plasminogen Kringle 5 Monoclonal antibody MAB742 Specifically detects Plasminogen Kringle 5 in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Plasminogen Kringle 5
120304
Western Blot 1 ug/mL
AAA50168
Plg
E. coli-derived recombinant mouse Plasminogen Kringle 5 Cys481-Ala563 Accession # AAA50168
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5340
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Plasminogen Kringle 5 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Plasminogen Kringle 5 is observed.