Learn More
Plasmodium falciparum HRP-2 Monoclonal Antibody (D3A), Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA518246
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes histidine rich protein II (HRP2) of P. falciparum. Purity is >90%. Recommended pairs for ELISA: MA5-18246 (capture) and MA5-18243 (conjugated, detection).
Plasmodium falciparum HRP-2 Monoclonal antibody specifically detects Plasmodium falciparum HRP-2 in Protozoa samples. It is validated for ELISA.Spezifikation
Plasmodium falciparum HRP-2 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG2b |
ELISA | |
D3A | |
Unconjugated | |
P. falciparum; P. falciparum HRP2; P. falciparum HRP-2 | |
Plasmodium falciparum recombinant protein | |
100 μg | |
Primary | |
4° C | |
Liquid |