missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR171 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA132573
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-32573 detects Platelet activating receptor H964 from human samples.
GPR171 Polyclonal antibody specifically detects GPR171 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
GPR171 | |
Polyclonal | |
Unconjugated | |
GPR171 | |
A630006E05; F730001G15Rik; G protein-coupled receptor 171; Gpr171; G-protein coupled receptor H963; H963; platelet activating receptor homolog; probable G-protein coupled receptor 171 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O14626 | |
GPR171 | |
Synthetic 16 amino acid peptide from 2nd extracellular domain of human GPR171 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |