Learn More
PLCG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529277
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Raji, Raw264.7. Predicted reactivity: Mouse (94%), Rat (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLCG2 Polyclonal antibody specifically detects PLCG2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLCG2 | |
Polyclonal | |
Unconjugated | |
Plcg2 | |
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2; APLAID; FCAS3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-gamma-2; phospholipase C gamma 2; phospholipase C, gamma 2; phospholipase C, gamma 2 (phosphatidylinositol-specific); phospholipase C-gamma-2; Phospholipase C-IV; PLC gamma 2; PLC gamma2; PLC- gamma2; PLC gamma-2; PLCG2; Plcg-2; PLC-gamma; PLCgamma2; PLC-gamma-2; PLC-IV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
234779, 5336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.75 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P16885, Q8CIH5 | |
Plcg2 | |
Recombinant fragment corresponding to a region within amino acids 70 and 360 of Human PLCG2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |