missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPAT9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538698
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This gene encodes a member of the lysophosphatidic acid acyltransferase protein family. The encoded protein is an enzyme which catalyzes the conversion of glycerol-3-phosphate to lysophosphatidic acid in the synthesis of triacylglycerol. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2012].Spezifikation
AGPAT9 | |
Polyclonal | |
Unconjugated | |
GPAT3 | |
GPAT3, MAG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
231510, 305166, 84803 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q53EU6, Q8C0N2, Q4V8J4 | |
GPAT3 | |
A synthetic peptide derived from the C-terminal region of human PLCH | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |