Learn More
PLCL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531543
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, U87-MG. Predicted reactivity: Mouse (98%), Rat (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLCL2 Polyclonal antibody specifically detects PLCL2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLCL2 | |
Polyclonal | |
Unconjugated | |
PLCL2 | |
inactive phospholipase C-like protein 2; KIAA1092; mKIAA1092; phospholipase C; phospholipase C like 2; phospholipase C, epsilon 2; phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLCE2; PLC-epsilon-2; PLC-L(2); Plcl2; PLC-L2; PRIP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UPR0 | |
PLCL2 | |
Recombinant protein fragment corresponding to a region within amino acids 92 and 358 of Human PLCL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |