missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523335
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PLD2 Polyclonal antibody specifically detects PLD2 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLD2 | |
Polyclonal | |
Unconjugated | |
Pld2 | |
choline phosphatase 2; hPLD2; phosphatidylcholine-hydrolyzing phospholipase D2; phospholipase D gene 2; phospholipase D2; PLD 2; PLD1C; PLD2; Pldc; rPLD2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 50% glycerol and 0.025% sodium azide | |
O14939 | |
Pld2 | |
Within the range amino acids 199-230 of human PLD2 protein | |
RUO | |
5338 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |