missing translation for 'onlineSavingsMsg'
Learn More

PLEKHG4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA522039

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13240069

  • 470.00 € / 100 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PLEKHG4 Polyclonal antibody specifically detects PLEKHG4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PLEKHG4
Polyclonal
Unconjugated
PLEKHG4
4931414L13Rik; AI448583; ARHGEF44; mFLJ00068; PH domain-containing family G member 4; pleckstrin homology and RhoGEF domain containing G4; pleckstrin homology domain containing, family G (with RhoGef domain) member 4; Pleckstrin homology domain-containing family G member 4; PLEKHG4; PRTPHN1; Puratrophin-1; Purkinje cell atrophy associated protein 1; Purkinje cell atrophy-associated protein 1; RGD1310790; SCA4
Rabbit
Antigen affinity chromatography
RUO
25894
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q58EX7
PLEKHG4
Recombinant fragment corresponding to a region within amino acids 35 and 256 of PLEKHG4 (Uniprot ID#Q58EX7)
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers