Learn More
Plexin-A1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547612
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A2 and rmPlexin A3 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Plexin-A1 Polyclonal antibody specifically detects Plexin-A1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
Plexin-A1 | |
Polyclonal | |
Unconjugated | |
PLXNA1 | |
2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV | |
Goat | |
Antigen affinity chromatography | |
RUO | |
18844, 5361 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P70206, Q9UIW2 | |
PLXNA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Plexin A1 Ser28-Pro1242 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |