Learn More
PLXNB2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA547880
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 9% cross-reactivity with recombinant human Plexin B2 is observed, and less than 1% cross-reactivity with recombinant mouse Plexin B3 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
PLXNB2 Polyclonal antibody specifically detects PLXNB2 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
PLXNB2 | |
Polyclonal | |
Unconjugated | |
PLXNB2 | |
Debt; differentially expressed in brain tumors; dJ402G11.3; KIAA0315; MM1; MM1KIAA0315dJ402G11.3; Nbla00445; PLEXB2; plexin B2; plexin-B2; PLXNB2 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
140570 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
B2RXS4 | |
PLXNB2 | |
CHO-derived recombinant mouse Plexin B2 Leu20-Trp1029 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |