missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527303
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PGF Polyclonal antibody specifically detects PGF in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PGF | |
Polyclonal | |
Unconjugated | |
PGF | |
AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.09 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P49763 | |
PGF | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 141 of Human PlGF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |