missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539441
Weitere Details : Netto-Gewicht : 0.00900kg
Spezifikation
PLK5 | |
Polyclonal | |
Unconjugated | |
PLK5 | |
PLK-5, PLK5P, SgK384ps, inactive serine/threonine-protein kinase PLK5, polo-like kinase 5 pseudogene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126520 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q496M5 | |
PLK5 | |
A synthetic peptide derived from the internal region of human PLK5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |