Learn More
PLSCR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527590
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: H1299, mouse stomach. Predicted reactivity: Mouse (94%), Rat (94%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLSCR3 Polyclonal antibody specifically detects PLSCR3 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
PLSCR3 | |
Polyclonal | |
Unconjugated | |
Plscr3 | |
2210403O21Rik; 2610037N06Rik; Ca 2+ dependent phospholipid scramblase 3; ca(2+)-dependent phospholipid scramblase 3; ESTM3; LOW QUALITY PROTEIN: phospholipid scramblase 3; LOW QUALITY PROTEIN: phospholipid scramblase 3; phospholipid scramblase 3; phospholipid scramblase 3; PL scramblase 3; Pls3; Plscr3; Unknown (protein for MGC:138060); X83310 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57048, 70310 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.77 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9JIZ9, Q9NRY6 | |
Plscr3 | |
Recombinant fragment corresponding to a region within amino acids 48 and 288 of Human PLSCR3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |