missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXDC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531215
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLXDC2 Polyclonal antibody specifically detects PLXDC2 in Human samples. It is validated for Western Blot.Spezifikation
PLXDC2 | |
Polyclonal | |
Unconjugated | |
PLXDC2 | |
1200007L24Rik; 5430431D22Rik; AI646728; AU022916; AV231634; plexin domain containing 2; plexin domain-containing protein 2; PLXDC2; TEM7R; tumor endothelial marker 7 related; tumor endothelial marker 7-related protein; UNQ2514/PRO6003 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84898 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.95 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6UX71 | |
PLXDC2 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 218 of Human PLXDC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |