missing translation for 'onlineSavingsMsg'
Learn More

PMCA3 ATPase Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA1916

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11528601

  • 508.00 € / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-916 detects calcium pump of the plasma membrane 3 (PMCA3 ATPase) from rat, human and porcine tissues. PA1-916 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽127 kDa protein representing PMCA3a and PMCA3b ATPase from rat brain microsomal fractions. PA1-916 immunizing peptide corresponds to amino acid residues 5-22 from rat PMCA3 ATPase protein. This sequence is completely conserved between rat and human PMCA3 ATPase.

PMCA3 ATPase Polyclonal antibody specifically detects PMCA3 ATPase in Human, Porcine, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

PMCA3 ATPase
Polyclonal
Unconjugated
ATP2B3
6430519O13Rik; ATP2B3; ATPase plasma membrane Ca2+ transporting 3; ATPase, Ca++ transporting, plasma membrane 3; CFAP39; cilia and flagella associated protein 39; CLA2; LOW QUALITY PROTEIN: plasma membrane calcium-transporting ATPase 3; OPCA; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 3; plasma membrane calcium pump; Plasma membrane calcium pump isoform 3; Plasma membrane calcium-transporting ATPase 3; PMCA3; PMCA3a; SCAX1
Rabbit
Antigen affinity chromatography
RUO
100520061, 29599, 492
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q16720, Q64568
ATP2B3
Synthetic Peptide: A(5) N S S I E F H P K P Q Q Q R E V P(22)
100 μg
Primary
Human, Porcine, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote