missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMPCA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546591
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: IRNVKPEDVK RVASKMLRGK PAVAALGDLT DLPTYEHIQT ALSSKDGRLP Sequence homology: Cow: 86%; Dog: 77%; Guinea Pig: 79%; Horse: 92%; Human: 100%; Mouse: 93%; Rat: 93%
PMPCA Polyclonal antibody specifically detects PMPCA in Human samples. It is validated for Western Blot.Spezifikation
PMPCA | |
Polyclonal | |
Unconjugated | |
PMPCA | |
1200002L24Rik; 4933435E07Rik; alpha-MPP; inactive zinc metalloprotease alpha; inositol polyphosphate-5-phosphatase, 72 kD; INPP5E; KIAA0123; mitochondrial matrix processing protease, alpha subunit; mitochondrial processing peptidase alpha subunit, mitochondrial; mitochondrial-processing peptidase subunit alpha; MPPA; P-55; peptidase (mitochondrial processing) alpha; peptidase, mitochondrial processing alpha subunit; peptidase, mitochondrial processing subunit alpha; PMPCA; SCAR2; Unknown (protein for MGC:139568) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23203 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q10713 | |
PMPCA | |
Synthetic peptide directed towards the C-terminal of human PMPCA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |