missing translation for 'onlineSavingsMsg'
Learn More

PMS2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA529835

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13494147

  • 472.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: A431, HeLa, HepG2, human PMS2-transfected 293T. Predicted reactivity: Mouse (88%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PMS2 Polyclonal antibody specifically detects PMS2 in Human samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

PMS2
Polyclonal
Unconjugated
PMS2
AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6
Rabbit
Antigen affinity chromatography
RUO
5395
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P54278
PMS2
Recombinant fragment corresponding to a region within amino acids 1 and 317 of Human PMS2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote