Learn More
anti-PPAR delta/NR1C2, Clone: K9436, R&D Systems™
Marke:  RD Systems (Bio-Techne) PP-K9436-10
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412466
  • 504.42 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Applications:
Direct ELISA, Gel Supershift, Western Blotting
Spezifikation
Spezifikation
PPAR delta/NR1C2
K9436
Western Blot, Direct ELISA, Gel Supershift Assay
Purified
PPARD
IgG2a
100 ug
Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Monoclonal
Human
Western Blot, ELISA, Gel Shift
Unconjugated
A liquid formulation in physiologic saline with 0.1% NaN3. with 0.1% Sodium Azide
FAAR, MGC3931, NR1C2, NR1C2PPAR-delta, NUC1, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB, PPARD
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5467
Videos