missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG Fc Cross-Adsorbed Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A16128
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rabbit IgG Fc Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG Fc Cross-Adsorbed in Rabbit samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
Rabbit IgG Fc Cross-Adsorbed | |
Polyclonal | |
Biotin | |
Goat | |
Affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobin Fc region | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |