Learn More
F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen PA533282
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33282 targets Rat IgG in FACS applications. Cross-reactivity with mouse IgG has been reduced by adsorption. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Rat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Cross-Adsorbed in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry.Spezifikation
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
FITC | |
Goat | |
affinity chromatography | |
RUO | |
Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
700 μg/mL | |
PBS with 0.5% BSA and 0.09% sodium azide | |
Rat IgG | |
500 μL | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |