Learn More
F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, PE, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen PA533283
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
F(ab')2 fragments were prepared by pepsin digestion. Cross-reactivity with mouse IgG has been reduced by adsorption. For FACS analysis, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Reconstitute with 0.5 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark.
Rat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Cross-Adsorbed in Rat samples. It is validated for Flow Cytometry.Spezifikation
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
PE | |
Goat | |
purified | |
RUO | |
Rat | |
Lyophilized |
Flow Cytometry | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
whole rat IgG | |
100 Tests | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |