missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Cyanine3, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A10522
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
Rat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Cross-Adsorbed in Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry.Spezifikation
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Cyanine3 | |
Goat | |
purified | |
RUO | |
Rat | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |