Learn More
Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A18876
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. 1 year from date of receipt. Prepare working dilution prior to use and then discard. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rat serum proteins, IgG from human or mouse, highly cross absorbed against mouse IgG.
Rat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Cross-Adsorbed in Rat samples. It is validated for Immunocytochemistry.Spezifikation
Rat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
TRITC | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Lyophilized |
Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |