Learn More
Mouse IgG1 Isotype Control, Biotin, Invitrogen™
Mouse Monoclonal Isotype Control
Marke: Invitrogen MA133298
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-33298 detects IgG1 from rat samples. MA1-33298 has been successfully used in immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue) applications.
Mouse IgG1 Monoclonal antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG1 | |
Monoclonal | |
Biotin | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Proprietary immunogen - Contact ABR for details. | |
100 Tests | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Control, Flow Cytometry | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |