missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2a Isotype Control, Invitrogen™

Rat Isotype Control

Marke:  Invitrogen 029688

136.00 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.50000kg

Artikelnummer. 10600854

  • 204.00 € / 1 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Spezifikation

Spezifikation

Rat IgG2a
1mg/mL
PBS with 0.1% sodium azide; pH 7.4
Rat
Purified
RUO
None
Liquid
Flow Cytometry
Unconjugated
IgG2, IgG2a
Rat IgG2a
500 μg
Isotype Control (Primary)
Store at -20°C, Avoid Freeze/Thaw Cycles
IgG2a
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote