Learn More
Rat IgG2a Isotype Control, Invitrogen™
Rat Isotype Control
Marke: Invitrogen PA533214
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33214 has been shown to be negative on all mouse cells and cell lines tested. This antibody recognizes a human cell surface marker, and therefore cannot be used as a negative control in this species. Test results have shown that PA5-33214 is also suitable for use as a negative control with canine tissues. This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Rat IgG2a Control antibody specifically detects Rat IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Rat IgG2a | |
1 mg/mL | |
PBS ohne Konservierungsmittel | |
Rat | |
Protein G | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Human lymphocytes | |
500 μg | |
Isotype Control (Primary) | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |