Learn More
Rat IgG2a Isotype Control, PE, Invitrogen™
Rat Isotype Control
Marke: Invitrogen PA533216
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-33216 has been shown to be negative on all mouse cells and cell lines tested. This antibody recognizes a human cell surface marker, and therefore cannot be used as a negative control in this species. Test results have shown that PA5-33216 is also suitable for use as a negative control with canine tissues. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Rat IgG2a Control antibody specifically detects Rat IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Rat IgG2a | |
Conc. Not Determined | |
PBS mit 1 % BSA, 5 % Saccharose und 0.09 % Natriumazid | |
Rat | |
Protein G | |
RUO | |
Not Applicable | |
Lyophilized |
Control, Flow Cytometry | |
PE | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Human lymphocytes. | |
100 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a |