Learn More
Mouse anti-Rat IgG2a (Heavy chain) Secondary Antibody, Biotin, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen MA516817
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse anti Rat IgG2a heavy chain antibody, clone MARG2a-1 recognizes the rat gamma 2a immunoglobulin heavy chain, and does not cross-react with other immunoglobulin classes or subclasses.
Rat IgG2a (Heavy chain) Monoclonal antibody specifically detects Rat IgG2a (Heavy chain) in Rat samples. It is validated for ELISA.Spezifikation
Rat IgG2a (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide | |
Rat IgG2a | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA | |
MARG2a-1 | |
Biotin | |
Mouse | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |