missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSK1 Monoclonal Antibody (257R21), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen AHO1472
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
RSK1 Monoclonal antibody specifically detects RSK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
RSK1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P18653, Q15418, Q63531 | |
RPS6KA1 | |
Recombinant fragment of human RSK1 expressed in E. coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Western Blot | |
257R21 | |
Unconjugated | |
RPS6KA1 | |
90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAPK-1; mapkapk1a; MAPKAPK-1a; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinas | |
Mouse | |
Affinity chromatography | |
RUO | |
20111, 6195, 81771 | |
-20°C | |
Liquid |