missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Monoclonal Antibody (SP127), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA516396
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is colon or melanoma tissue.
S100B Monoclonal antibody specifically detects S100B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).Spezifikation
S100B | |
Monoclonal | |
0.22 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P04271 | |
S100B | |
Full length recombinant human S100 beta. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
SP127 | |
Unconjugated | |
S100B | |
AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P | |
Rabbit | |
Protein A/G | |
RUO | |
6285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |