Learn More
S100 Monoclonal Antibody (4C4.9), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA512966
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-12966 targets S100 Protein in IHC (P) applications and shows reactivity with Bovine, Human, mouse, and Rat samples. The MA5-12966 immunogen is purified bovine brain S100 protein.
S100 Monoclonal antibody specifically detects S100 in Bovine, Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
S100 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02638, P02639, P04271, P23297, P50114, P56565 | |
S100A1, S100B | |
Purified bovine brain S100 protein | |
500 μL | |
Primary | |
Bovine, Human, Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin) | |
4C4.9 | |
Biotin | |
S100A1 | |
AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
Mouse | |
Protein A | |
RUO | |
20193, 20203, 525716, 528735, 6271, 6285 | |
4° C | |
Liquid |