Learn More
S100A1 Monoclonal Antibody (2C8B8, 2C8E8,3C8D8), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515325
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15325 targets S100A1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15325 immunogen is purified recombinant fragment of S100A1 expressed in E. Coli.
S100A1 Monoclonal antibody specifically detects S100A1 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
S100A1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P23297 | |
S100A1 | |
Purified recombinant fragment of S100A1 expressed in E. Coli. | |
RUO | |
6271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
2C8B8, 2C8E8,3C8D8, 2C8E8, 3C8D8 | |
Unconjugated | |
S100A1 | |
AI266795; Protein S100-A1; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium-binding protein A1; S-100 protein alpha chain; S-100 protein subunit alpha; S100 protein, alpha polypeptide; S100a; S100A1; S100-alpha | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |