missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A10 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA175390
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-75390 specifically detects S100A10. PA1-75390 has been successfully used in ELISA applications. The PA1-75390 immunogen is Recombinant S100A10.
S100A10 Polyclonal antibody specifically detects S100A10 in Human samples. It is validated for ELISA.Spezifikation
S100A10 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P60903 | |
S100A10 | |
Recombinant human S100A10 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
S100A10 | |
42C; AA409961; AL024248; Annexin II; annexin II ligand; annexin II ligand, calpactin I, light polypeptide; annexin II tetramer (AIIt) p11 subunit; ANX2L; ANX2LG; Ca[1]; CAL12; Cal1l; calcium binding protein A11 (calgizzarin); calpactin I light chain; Calpactin-1 light chain; cellular ligand of annexin II; CLP11; GP11; MGC111133; MGC133268; Nerve growth factor-induced protein 42C; OTTHUMP00000015270; P PRSS26; p10; p10 protein; P11; p11 subunit; Protein S100 A10; Protein S100-A10; S100 calcium binding protein A10; S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S100 calcium binding protein A10 (calgizzarin); S100 calcium binding protein A10 (calpactin); S100 calcium-binding protein A10; S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S-100 related protein, clone 42C; S100a10 | |
Sheep | |
Affinity chromatography | |
RUO | |
6281 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |