missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A12 Monoclonal Antibody (161205), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523925
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody detects recombinant human EN-RAGE in direct ELISAs. Reconstitute at 0.5 mg/mL in sterile PBS.
S100A12 Monoclonal antibody specifically detects S100A12 in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
S100A12 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P80511 | |
S100A12 | |
E. coli-derived recombinant human EN-RAGE Met1-Glu92 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
161205 | |
Unconjugated | |
S100A12 | |
CAAF1; CAGC; Calcitermin; calcium-binding protein in amniotic fluid 1; calgranulin C; calgranulin-C; CGRP; EN RAGE; ENRAGE; EN-RAGE; Extracellular newly identified RAGE-binding protein; migration inhibitory factor-related protein 6; MRP6; MRP-6; Neutrophil S100 protein; p6; Protein S100 A12; protein S100-A12; S100 calcium binding protein A12; S100 calcium-binding protein A12; S100 calcium-binding protein A12 (calgranulin C); S100A12 | |
Mouse | |
Protein A/G | |
RUO | |
6283 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |