Learn More
S100A16 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA547840
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 30% cross-reactivity with recombinant human S100A16 is observed, and less than 1% cross-reactivity with recombinant mouse S100A4, recombinant human (rh) S100A2, and rhS100A10 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
S100A16 Polyclonal antibody specifically detects S100A16 in Mouse samples. It is validated for Western Blot.Spezifikation
S100A16 | |
Polyclonal | |
Unconjugated | |
S100a16 | |
2300002L21Rik; AAG13; aging-associated gene 13 protein; aging-associated protein 13; AI325039; AI663996; DT1P1A7; MGC17528; protein S100-A16; Protein S100F; Protein S100-F; S100 calcium binding protein A16; S100 calcium-binding protein A16; S100A16; S100F | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
67860 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9D708 | |
S100a16 | |
E. coli-derived recombinant mouse S100A16 Ala2-Ser124 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |