missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531861
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431. Predicted reactivity: Dog (86%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
S100A2 Polyclonal antibody specifically detects S100A2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
S100A2 | |
Polyclonal | |
Unconjugated | |
S100A2 | |
CAN19; EG628324; predicted gene, EG628324; Protein S100-A2; Protein S-100L; S100 calcium binding protein A2; S100 calcium-binding protein A2; S100a2; S100L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.85 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P29034 | |
S100A2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 98 of Human S100A2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |