missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539372
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown. [provided by RefSeq, Jul 2008].Spezifikation
S100A3 | |
Polyclonal | |
Unconjugated | |
S100A3 | |
S100E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114216, 20197, 6274 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P33764, P62818, P62819 | |
S100A3 | |
A synthetic peptide derived from the internal region of human S100A3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |