missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518601
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
S100A4 Polyclonal antibody specifically detects S100A4 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
S100A4 | |
Polyclonal | |
Unconjugated | |
S100a4 | |
18A2; 42a; calvasculin; CAPL; fibroblast-specific protein-1; FSP1; leukemia multidrug resistance associated protein; malignant transformation suppression 1; metastasin; Metastatic cell protein; Mts1; Nerve growth factor-induced protein 42A; P9K; P9KA; PeL98; pk9a; Placental calcium-binding protein; pr; protein 18A2; Protein 9 Ka homologous to calcium-binding protein; protein Mts1; Protein S100-A4; RNP9KA; S100 calcium binding protein A4; S100 calcium-binding protein A4; S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog); S100a4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P26447 | |
S100a4 | |
Synthetic peptide sequence (CNEFFEGFPDKQP) corresponding to the C-terminus amino acids of S100A4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |