missing translation for 'onlineSavingsMsg'
Learn More

S100A8 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA547439

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15537932

  • 412.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) S100A9, rmS100A10, and recombinant human S100B is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

S100A8 Polyclonal antibody specifically detects S100A8 in Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

S100A8
Polyclonal
Unconjugated
S100A8
60B8AG; AI323541; B8Ag; CAGA; CAGB; calgranulin A; calgranulin B; calgranulin-A; Calgranulin-B; calprotectin L1H subunit; Calprotectin L1L subunit; CFAg; CGLA; CGLB; chemotactic cytokine CP-10; chemotactic S100 protein; CP-10; Cystic fibrosis antigen; L1Ag; leukocyte L1 complex heavy chain; Leukocyte L1 complex light chain; LIAG; MA387; MAC387; MIF; Migration inhibitory factor-related protein 14; migration inhibitory factor-related protein 8; MRP14; MRP-14; MRP8; MRP-8; NIF; P14; p8; Pro-inflammatory S100 cytokine; protein S100-A8; Protein S100-A9; S100 A8; S100 calcium binding protein A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium binding protein A9; S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100A8; S100A9; Urinary stone protein band A
Goat
Antigen affinity chromatography
RUO
116547, 20201
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunocytochemistry, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P27005, P50115
S100A8
E. coli-derived recombinant mouse S100A8 Pro2-Glu89
100 μg
Primary
Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote