missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Monoclonal Antibody (357517), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524106
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) S100A10 or rhS100B is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
S100P | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P25815 | |
S100P | |
E. coli-derived recombinant human S100P Met1-Lys95 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
357517 | |
Unconjugated | |
S100P | |
MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100p | |
Mouse | |
Protein A/G | |
RUO | |
6286 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |