Learn More
S100PBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544508
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ERRLGKVIPV LQTKTRTNVP TFSQSNLEQQ KQLYLRSVIA HIEDPEDTNQ Sequence homology: Cow: 85%; Dog: 85%; Horse: 86%; Human: 100%; Mouse: 85%; Pig: 85%; Rabbit: 77%; Rat: 85%
S100PBP Polyclonal antibody specifically detects S100PBP in Human samples. It is validated for Western Blot.Spezifikation
S100PBP | |
Polyclonal | |
Unconjugated | |
S100PBP | |
4930429A08Rik; AI851343; hypothetical protein LOC539983; RGD1564943; S100P binding protein; S100P binding protein 1; S100P-binding protein; S100P-binding protein Riken; S100pbp; S100PBPR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64766 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96BU1 | |
S100PBP | |
Synthetic peptide directed towards the middle region of human S100PBP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |