missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1PR2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519363
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine based on sequence homology.
S1PR2 Polyclonal antibody specifically detects S1PR2 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
S1PR2 | |
Polyclonal | |
Unconjugated | |
S1PR2 | |
agr16; edg5; edg-5; S1PR2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14739, 9294 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O95136, P52592 | |
S1PR2 | |
Synthetic peptide sequence (NYTKETLETQ) corresponding to the internal amino acids of S1PR2 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |