missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S6 Monoclonal Antibody (C.896.4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515123
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
S6 Monoclonal antibody specifically detects S6 in Drosophila, Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
S6 | |
Monoclonal | |
4 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P29327, P62753, P62754, P62755 | |
RPS6 | |
Recombinant fusion protein corresponding to full-length human S6 ribosomal protein | |
100 μL | |
Primary | |
Drosophila, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
C.896.4 | |
Unconjugated | |
RPS6 | |
(Rp)S6; 40S ribosomal protein S6; air[8]; air8; anon-WO02059370.61; CG10944; CG10944-PA; CG10944-PB; Dmel\CG10944; Dmel_CG10944; DS6; hemocytes enlarged; hen; l(1)air[8]; l(1)air8; l(1)hen; LD31286p; lethal(1)aberrant immune response[8]; lethal-(1)-haemocytes enlarged; lethal-aberrant-immune-response-8; M(1)7BC; M(1)7C; minute; minute (1) 7BC; OK/SW-cl.2; OTTHUMP00000021121; OTTHUMP00000021122; Phosphoprotein NP33; pp30; ribosomal protein S6; Rp S6; RP11-513M16.6; RPS6; RpS6-PA; RpS6-PB; RS6; S6; S6 ribosomal protein; S6R; Small ribosomal subunit protein eS6; wu:fa92e06; wu:fb64g06; zgc:92237 | |
Mouse | |
Affinity chromatography | |
RUO | |
20104, 29304, 31700, 6194 | |
-20°C | |
Liquid |