missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538824
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
The family of ribosomal S6 kinases (Rsks), designated Rsk-1 (MAPKAP kinase-1), Rsk-2 and Rsk-3, are intracellular serine/threonine kinases that are important signaling intermediates in response to a broad range of ligand activated receptor tyrosine kinases. A unique feature common to the members of the Rsk family is that each possesses two non-identical complete kinase catalytic domains. An additional Rsk protein, Rsk-4, shows a high level of homology to the three previously isolated members of the human Rsk family. Rsk-4 is most abundantly expressed in brain and kidney and plays a role in normal neuronal development. The 70 kDa family of ribosomal S6 kinases includes p70 S6 kinase and p70 S6 kinase beta, which are thought to have similar regulatory functions. MSK1 (also designated RLPK) is a novel Rskrelated protein, which, like the p90 Rsk family members, contains two non-identical complete kinase catalytic domains.Spezifikation
RSK4 | |
Polyclonal | |
Unconjugated | |
RPS6KA6 | |
Ribosomal protein S6 kinase alpha-6, S6K-alpha-6, 90 kDa ribosomal protein S6 kinase 6, p90-RSK 6, p90RSK6, Ribosomal S6 kinase 4, RSK-4, pp90RSK4, RPS6KA6, RSK4, S6KA6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27330 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UK32 | |
RPS6KA6 | |
A synthetic peptide derived from the internal region of human S6K-alpha6 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |