Learn More
RPS6KB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527561
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RPS6KB2 Polyclonal antibody specifically detects RPS6KB2 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
RPS6KB2 | |
Polyclonal | |
Unconjugated | |
Rps6kb2 | |
70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; RPS6KB2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14B; SRK; STK14B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58988, 6199 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9UBS0, Q9Z1M4 | |
Rps6kb2 | |
Recombinant protein fragment corresponding to a region within amino acids 208 and 441 of Human S6K2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |