missing translation for 'onlineSavingsMsg'
Learn More

SQSTM1 Recombinant Rabbit Monoclonal Antibody (11HC14LC25), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15377134

  • 469.00 € / 100 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with bovine, feline, non-human primate, mouse, rat, ovine and porcine based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

SQSTM1 Recombinant Monoclonal antibody specifically detects SQSTM1 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

SQSTM1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3
proprietary
100 μg
Primary
Human
Liquid
ELISA, Immunocytochemistry, Western Blot
11HC14LC25
Unconjugated
Q13501
Rabbit
Protein A
RUO
8878
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote