Learn More
Serpin C1/Antithrombin-III Rat anti-Mouse, Clone: 208730, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1287
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418525
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 04-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Serpin C1/Antithrombin-III Monoclonal antibody MAB1287 Specifically detects Serpin C1/Antithrombin-III in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Serpin C1/Antithrombin-III
208730
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINC1
Mouse myeloma cell line NS0-derived recombinant mouse Serpin C1/Antithrombin-III His33-Asn465 Accession # P32261
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
462
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P32261
Antithrombin-III, AT3antithrombin-III, ATIII, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Serpin C1/Antithrombin-III in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human (rh) Serpin C1 is observed and no cross-reactivity with rhSerpin A1, A3, F2, recombinant mouse Serpin D1 or A5 is observed.